The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000660.5 | NP_000651.3 | 2 - 3 | 1406 | 89 |
XM_011527242.1 | XP_011525544.1 | 2 - 3 | 1399 | 89 | |
GenBank mRNA | AK291907.1 | - | 2 - 3 | 895 | 89 |
AK307742.1 | - | 2 - 3 | 1091 | 89 | |
BC000125.1 | - | 2 - 3 | 963 | 89 | |
BC001180.1 | - | 2 - 3 | 963 | 89 | |
BC022242.1 | - | 2 - 3 | 886 | 89 | |
BT007245.1 | - | 2 - 3 | 517 | 89 |