Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLH3
Entrez Gene ID
27030
Gene Name
mutL homolog 3
Gene Aliases
HNPCC7
Chromosome Location
Chr.14: 75013764 - 75051532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014381.2 NP_055196.2 6 - 7 3861 75
XM_005267533.4 XP_005267590.1 5 - 6 3756 75
XM_017021219.1 XP_016876708.1 6 - 7 3787 75
GenBank mRNA AF195657.1 - 6 - 7 3759 75
BC036866.2 - 4 - 5 365 75
BC112167.1 - 5 - 6 3681 75

View Full Product Details