Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLH3
Entrez Gene ID
27030
Gene Name
mutL homolog 3
Gene Aliases
HNPCC7
Chromosome Location
Chr.14: 75013764 - 75051532 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040108.1 NP_001035197.1 6 - 7 3857 80
XM_005267532.4 XP_005267589.1 5 - 6 3752 80
XM_005267534.2 XP_005267591.1 6 - 7 3857 80
XM_006720116.3 XP_006720179.1 6 - 7 3769 80
GenBank mRNA AB039667.1 - 5 - 6 3641 80
AL833875.1 - 5 - 6 1687 80

View Full Product Details