The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024524.3 | NP_078800.3 | 12 - 13 | 1716 | 130 |
XM_005269357.3 | XP_005269414.1 | 12 - 13 | 1737 | 130 | |
XM_011513120.2 | XP_011511422.1 | 13 - 14 | 1838 | 130 | |
XM_011513121.1 | XP_011511423.1 | 12 - 13 | 1716 | 130 | |
XM_011513122.1 | XP_011511424.1 | 12 - 13 | 1410 | 130 | |
XM_011513123.2 | XP_011511425.1 | 13 - 14 | 1843 | 130 | |
XM_011513124.2 | XP_011511426.1 | 13 - 14 | 1843 | 130 | |
GenBank mRNA | AJ306929.1 | - | 3 - 4 | 2240 | 130 |
AK024639.1 | - | 1 - 2 | 113 | 130 | |
AK225064.1 | - | 1 - 2 | 113 | 130 | |
DR005713.1 | - | 5 - 6 | 597 | 130 |