The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024524.3 | NP_078800.3 | 23 - 24 | 3030 | 76 |
XM_005269357.3 | XP_005269414.1 | 23 - 24 | 3051 | 76 | |
XM_011513120.2 | XP_011511422.1 | 24 - 25 | 3152 | 76 | |
XM_011513121.1 | XP_011511423.1 | 23 - 24 | 3030 | 76 | |
XM_011513122.1 | XP_011511424.1 | 23 - 24 | 2724 | 76 | |
XM_011513123.2 | XP_011511425.1 | 24 - 25 | 3157 | 76 | |
XM_011513124.2 | XP_011511426.1 | 24 - 25 | 3157 | 76 | |
GenBank mRNA | AJ306929.1 | - | 14 - 15 | 3554 | 76 |
AK024639.1 | - | 12 - 13 | 1427 | 76 | |
AK075094.1 | - | 4 - 5 | 475 | 76 | |
AK225064.1 | - | 12 - 13 | 1427 | 76 |