The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024524.3 | NP_078800.3 | 26 - 27 | 3268 | 117 |
XM_005269357.3 | XP_005269414.1 | 26 - 27 | 3289 | 117 | |
XM_011513120.2 | XP_011511422.1 | 27 - 28 | 3390 | 117 | |
XM_011513121.1 | XP_011511423.1 | 26 - 27 | 3268 | 117 | |
XM_011513122.1 | XP_011511424.1 | 26 - 27 | 2962 | 117 | |
XM_011513123.2 | XP_011511425.1 | 27 - 28 | 3395 | 117 | |
XM_011513124.2 | XP_011511426.1 | 27 - 28 | 3395 | 117 | |
GenBank mRNA | AK024639.1 | - | 15 - 16 | 1665 | 117 |
AK075094.1 | - | 7 - 8 | 713 | 117 | |
AK225064.1 | - | 15 - 16 | 1665 | 117 |