The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024524.3 | NP_078800.3 | 4 - 5 | 810 | 145 |
XM_005269357.3 | XP_005269414.1 | 4 - 5 | 831 | 145 | |
XM_011513120.2 | XP_011511422.1 | 5 - 6 | 932 | 145 | |
XM_011513121.1 | XP_011511423.1 | 4 - 5 | 810 | 145 | |
XM_011513123.2 | XP_011511425.1 | 5 - 6 | 937 | 145 | |
XM_011513124.2 | XP_011511426.1 | 5 - 6 | 937 | 145 | |
GenBank mRNA | DV080200.1 | - | 3 - 4 | 145 |