The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003652.3 | NP_001003652.1 | 10 - 11 | 1677 | 119 |
NM_001135937.2 | NP_001129409.1 | 9 - 10 | 1587 | 119 | |
NM_005901.5 | NP_005892.1 | 10 - 11 | 1569 | 119 | |
XM_005258259.3 | XP_005258316.1 | 11 - 12 | 1810 | 119 | |
XM_006722451.3 | XP_006722514.1 | 11 - 12 | 1822 | 119 | |
XM_011525984.2 | XP_011524286.1 | 10 - 11 | 1166 | 119 | |
XM_011525985.2 | XP_011524287.1 | 12 - 13 | 1869 | 119 | |
XM_017025745.1 | XP_016881234.1 | 12 - 13 | 1908 | 119 | |
XM_017025746.1 | XP_016881235.1 | 10 - 11 | 1720 | 119 | |
XM_017025747.1 | XP_016881236.1 | 11 - 12 | 1795 | 119 | |
XM_017025748.1 | XP_016881237.1 | 12 - 13 | 1881 | 119 | |
XM_017025750.1 | XP_016881239.1 | 11 - 12 | 1770 | 119 | |
GenBank mRNA | AF027964.1 | - | 10 - 11 | 1383 | 119 |
AK312963.1 | - | 9 - 10 | 1406 | 119 | |
BC014840.1 | - | 10 - 11 | 1525 | 119 | |
BC025699.1 | - | 10 - 11 | 1383 | 119 | |
BT007422.1 | - | 9 - 10 | 1277 | 119 | |
U59911.1 | - | 10 - 11 | 1376 | 119 | |
U65019.1 | - | 9 - 10 | 1313 | 119 | |
U68018.1 | - | 9 - 10 | 1332 | 119 |