Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC14A1
Entrez Gene ID
6563
Gene Name
solute carrier family 14 member 1 (Kidd blood group)
Gene Aliases
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE
Chromosome Location
Chr.18: 45724123 - 45752520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128588.3 NP_001122060.3 5 - 6 734 59
NM_001146036.2 NP_001139508.2 5 - 6 663 59
NM_001146037.1 NP_001139509.1 3 - 4 831 59
NM_001308278.1 NP_001295207.1 3 - 4 408 59
NM_015865.6 NP_056949.4 4 - 5 576 59
XM_005258329.1 XP_005258386.1 5 - 6 710 59
XM_006722526.3 XP_006722589.1 5 - 6 711 59
XM_011526142.1 XP_011524444.1 4 - 5 1491 59
XM_011526144.1 XP_011524446.1 5 - 6 710 59
GenBank mRNA AK091064.1 - 3 - 4 1059 59
AK123681.1 - 2 - 3 1635 59
AK289608.1 - 5 - 6 677 59
AK294129.1 - 3 - 4 347 59
BC050539.1 - 4 - 5 502 59
GQ502682.1 - 2 - 3 365 59
HQ709264.1 - 2 - 3 344 59
KC677694.1 - 2 - 3 344 59
L36121.1 - 2 - 3 362 59
U35735.1 - 4 - 5 512 59
Y19039.1 - 2 - 3 365 59

View Full Product Details