Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC14A1
Entrez Gene ID
6563
Gene Name
solute carrier family 14 member 1 (Kidd blood group)
Gene Aliases
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE
Chromosome Location
Chr.18: 45724123 - 45752520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128588.3 NP_001122060.3 6 - 7 864 105
NM_001146036.2 NP_001139508.2 6 - 7 793 105
NM_001146037.1 NP_001139509.1 4 - 5 961 105
NM_001308278.1 NP_001295207.1 4 - 5 538 105
NM_001308279.1 NP_001295208.1 3 - 4 348 105
NM_015865.6 NP_056949.4 5 - 6 706 105
XM_005258329.1 XP_005258386.1 6 - 7 840 105
XM_005258333.1 XP_005258390.1 2 - 3 254 105
XM_006722526.3 XP_006722589.1 6 - 7 841 105
XM_011526142.1 XP_011524444.1 5 - 6 1621 105
XM_011526144.1 XP_011524446.1 6 - 7 840 105
GenBank mRNA AF328892.1 - 3 - 4 264 105
AK091064.1 - 4 - 5 1189 105
AK123681.1 - 3 - 4 1765 105
AK127452.1 - 4 - 5 450 105
AK289608.1 - 6 - 7 807 105
AK294129.1 - 4 - 5 477 105
AK295268.1 - 3 - 4 287 105
BC050539.1 - 5 - 6 632 105
DA156178.1 - 3 - 4 348 107
GQ502682.1 - 3 - 4 495 105
HQ709264.1 - 3 - 4 474 105
KC677694.1 - 3 - 4 474 105
L36121.1 - 3 - 4 492 105
U35735.1 - 5 - 6 642 105
Y19039.1 - 3 - 4 495 105

View Full Product Details