The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004124.2 | NP_004115.1 | 2 - 3 | 188 | 99 |
GenBank mRNA | AB001106.1 | - | 2 - 3 | 186 | 99 |
AB209240.1 | - | 2 - 3 | 120 | 99 | |
AK311753.1 | - | 2 - 3 | 122 | 99 | |
BC005359.1 | - | 2 - 3 | 142 | 99 | |
BI547506.1 | - | 2 - 3 | 198 | 99 | |
BT006784.1 | - | 1 - 2 | 89 | 99 | |
CR542113.1 | - | 1 - 2 | 89 | 99 | |
M86492.1 | - | 2 - 3 | 129 | 99 |