The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_079834.2 | NP_524558.1 | 1 - 2 | 65 | 101 |
GenBank mRNA | AK074586.1 | - | 1 - 2 | 67 | 101 |