The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014786.3 | NP_055601.2 | 6 - 7 | 3854 | 85 |
XM_017018623.1 | XP_016874112.1 | 6 - 7 | 2245 | 85 | |
XM_017018624.1 | XP_016874113.1 | 5 - 6 | 1324 | 85 | |
GenBank mRNA | AB002335.1 | - | 6 - 7 | 2628 | 85 |
AF378754.1 | - | 6 - 7 | 3848 | 85 | |
BC137228.1 | - | 6 - 7 | 3854 | 85 |