The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014786.3 | NP_055601.2 | 8 - 9 | 4021 | 88 |
XM_017018623.1 | XP_016874112.1 | 8 - 9 | 2412 | 88 | |
XM_017018624.1 | XP_016874113.1 | 7 - 8 | 1491 | 88 | |
GenBank mRNA | AB002335.1 | - | 8 - 9 | 2795 | 88 |
AF378754.1 | - | 8 - 9 | 4015 | 88 | |
BC137228.1 | - | 8 - 9 | 4021 | 88 |