Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGIP1
Entrez Gene ID
84251
Gene Name
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 66533272 - 66751139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308203.1 NP_001295132.1 16 - 17 1361 90
NM_032291.3 NP_115667.2 19 - 20 2138 90
XM_005271264.3 XP_005271321.1 19 - 20 1972 90
XM_005271268.3 XP_005271325.1 17 - 18 1387 90
XM_005271270.4 XP_005271327.1 16 - 17 1694 90
XM_006710961.2 XP_006711024.1 19 - 20 1978 90
XM_006710966.2 XP_006711029.1 18 - 19 1912 90
XM_006710967.2 XP_006711030.1 18 - 19 1900 90
XM_006710969.2 XP_006711032.1 18 - 19 1888 90
XM_006710971.2 XP_006711034.1 18 - 19 1477 90
XM_006710972.2 XP_006711035.1 18 - 19 1471 90
XM_006710973.2 XP_006711036.1 16 - 17 1453 90
XM_006710974.2 XP_006711037.1 15 - 16 1369 90
XM_011542291.1 XP_011540593.1 18 - 19 1634 90
XM_011542292.1 XP_011540594.1 17 - 18 1574 90
XM_011542293.1 XP_011540595.1 14 - 15 1303 90
XM_017002505.1 XP_016857994.1 20 - 21 2026 90
XM_017002506.1 XP_016857995.1 21 - 22 2149 90
XM_017002507.1 XP_016857996.1 20 - 21 2201 90
XM_017002508.1 XP_016857997.1 21 - 22 2147 90
XM_017002509.1 XP_016857998.1 18 - 19 2002 90
XM_017002510.1 XP_016857999.1 19 - 20 2125 90
XM_017002511.1 XP_016858000.1 20 - 21 2083 90
XM_017002512.1 XP_016858001.1 20 - 21 1956 90
XM_017002513.1 XP_016858002.1 19 - 20 2135 90
XM_017002514.1 XP_016858003.1 19 - 20 1954 90
XM_017002515.1 XP_016858004.1 17 - 18 1930 90
XM_017002516.1 XP_016858005.1 18 - 19 2053 90
XM_017002517.1 XP_016858006.1 19 - 20 2022 90
XM_017002518.1 XP_016858007.1 19 - 20 2010 90
XM_017002519.1 XP_016858008.1 19 - 20 2035 90
XM_017002520.1 XP_016858009.1 17 - 18 1876 90
XM_017002521.1 XP_016858010.1 17 - 18 1828 90
XM_017002522.1 XP_016858011.1 18 - 19 1950 90
XM_017002523.1 XP_016858012.1 18 - 19 1974 90
XM_017002524.1 XP_016858013.1 18 - 19 1617 90
XM_017002525.1 XP_016858014.1 17 - 18 1556 90
XM_017002526.1 XP_016858015.1 16 - 17 1435 90
XM_017002527.1 XP_016858016.1 17 - 18 1459 90
XM_017002528.1 XP_016858017.1 17 - 18 1545 90
XM_017002529.1 XP_016858018.1 17 - 18 1411 90
XM_017002530.1 XP_016858019.1 16 - 17 1387 90
XM_017002531.1 XP_016858020.1 15 - 16 1387 90
XM_017002532.1 XP_016858021.1 16 - 17 1381 90
XM_017002533.1 XP_016858022.1 16 - 17 1489 90
XM_017002534.1 XP_016858023.1 15 - 16 1363 90
XM_017002535.1 XP_016858024.1 16 - 17 1477 90
XM_017002536.1 XP_016858025.1 16 - 17 1327 90
XM_017002537.1 XP_016858026.1 15 - 16 1416 90
GenBank mRNA AB210039.1 - 19 - 20 1920 90
AK090697.1 - 10 - 11 918 90
AK125044.1 - 2 - 3 905 90
AK298970.1 - 16 - 17 1324 90
AK299941.1 - 17 - 18 1398 90
AL136561.1 - 19 - 20 1812 90
BC040516.1 - 21 - 22 90
BX640813.1 - 16 - 17 1338 90
CR749541.1 - 16 - 17 1549 90

View Full Product Details