The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032291.3 | NP_115667.2 | 3 - 4 | 500 | 73 |
XM_006710967.2 | XP_006711030.1 | 3 - 4 | 322 | 73 | |
XM_017002505.1 | XP_016857994.1 | 3 - 4 | 322 | 73 | |
XM_017002506.1 | XP_016857995.1 | 4 - 5 | 445 | 73 | |
XM_017002509.1 | XP_016857998.1 | 3 - 4 | 322 | 73 | |
XM_017002510.1 | XP_016857999.1 | 4 - 5 | 445 | 73 | |
XM_017002511.1 | XP_016858000.1 | 4 - 5 | 445 | 73 | |
XM_017002512.1 | XP_016858001.1 | 4 - 5 | 318 | 73 | |
XM_017002517.1 | XP_016858006.1 | 4 - 5 | 444 | 73 | |
XM_017002524.1 | XP_016858013.1 | 4 - 5 | 438 | 73 | |
XM_017002525.1 | XP_016858014.1 | 4 - 5 | 437 | 73 | |
XM_017002526.1 | XP_016858015.1 | 3 - 4 | 322 | 73 | |
XM_017002527.1 | XP_016858016.1 | 4 - 5 | 346 | 73 | |
XM_017002530.1 | XP_016858019.1 | 3 - 4 | 322 | 73 | |
XM_017002533.1 | XP_016858022.1 | 4 - 5 | 436 | 73 | |
GenBank mRNA | AL136561.1 | - | 3 - 4 | 174 | 73 |
DA801276.1 | - | 3 - 4 | 315 | 73 |