The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016309.2 | NP_057393.2 | 6 - 7 | 728 | 84 |
XM_005255354.3 | XP_005255411.1 | 6 - 7 | 517 | 84 | |
XM_011545864.1 | XP_011544166.1 | 7 - 8 | 842 | 84 | |
GenBank mRNA | AF037601.1 | - | 6 - 7 | 604 | 84 |
AF151826.1 | - | 6 - 7 | 712 | 84 | |
AK025884.1 | - | 7 - 8 | 835 | 84 | |
AK223364.1 | - | 7 - 8 | 886 | 84 | |
AK225271.1 | - | 6 - 7 | 672 | 84 | |
AK291885.1 | - | 6 - 7 | 672 | 84 | |
AK314409.1 | - | 6 - 7 | 673 | 84 | |
AL137283.1 | - | 6 - 7 | 532 | 84 | |
BC001214.1 | - | 6 - 7 | 695 | 84 | |
BC014217.2 | - | 7 - 8 | 816 | 84 |