Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MLLT1
Entrez Gene ID
4298
Gene Name
MLLT1, super elongation complex subunit
Gene Aliases
ENL, LTG19, YEATS1
Chromosome Location
Chr.19: 6210381 - 6279948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005934.3 NP_005925.2 11 - 12 1715 76
XM_011528021.1 XP_011526323.1 11 - 12 1689 76
XM_011528022.1 XP_011526324.1 11 - 12 1572 76
XM_011528023.1 XP_011526325.1 10 - 11 1563 76
XM_017026819.1 XP_016882308.1 11 - 12 2271 76
GenBank mRNA D14539.1 - 11 - 12 1715 76
L04285.1 - 10 - 11 1551 76

View Full Product Details