Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MLLT1
Entrez Gene ID
4298
Gene Name
MLLT1, super elongation complex subunit
Gene Aliases
ENL, LTG19, YEATS1
Chromosome Location
Chr.19: 6210381 - 6279948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005934.3 NP_005925.2 8 - 9 1469 62
XM_011528021.1 XP_011526323.1 8 - 9 1443 62
XM_011528022.1 XP_011526324.1 8 - 9 1326 62
XM_011528023.1 XP_011526325.1 7 - 8 1317 62
XM_017026819.1 XP_016882308.1 8 - 9 2025 62
GenBank mRNA D14539.1 - 8 - 9 1469 62
L04285.1 - 7 - 8 1305 62

View Full Product Details