The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004352.3 | NP_004343.1 | 1 - 2 | 634 | 81 |
GenBank mRNA | AK314276.1 | - | 1 - 2 | 608 | 81 |
BC093692.1 | - | 2 - 3 | 312 | 81 | |
BC093718.1 | - | 2 - 3 | 312 | 81 | |
M58583.1 | - | 1 - 2 | 582 | 81 |