Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TARBP2
Entrez Gene ID
6895
Gene Name
TARBP2, RISC loading complex RNA binding subunit
Gene Aliases
LOQS, TRBP, TRBP1, TRBP2
Chromosome Location
Chr.12: 53500921 - 53506431 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134323.1 NP_599150.1 1 - 2 545 97
XM_005269117.1 XP_005269174.1 1 - 2 164 97
XM_005269120.4 XP_005269177.1 1 - 2 545 97
XM_017019911.1 XP_016875400.1 1 - 2 175 97
GenBank mRNA AK290737.1 - 1 - 2 118 97
AK293728.1 - 1 - 2 116 97
AK309569.1 - 1 - 2 164 97
BC002419.2 - 1 - 2 545 97
BC005860.2 - 1 - 2 410 97
BT007140.1 - 1 - 2 57 97
U08998.2 - 1 - 2 145 97

View Full Product Details