The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004178.4 | NP_004169.3 | 3 - 4 | 354 | 92 |
NM_134323.1 | NP_599150.1 | 3 - 4 | 819 | 92 | |
NM_134324.2 | NP_599151.2 | 3 - 4 | 394 | 92 | |
XM_005269114.1 | XP_005269171.1 | 3 - 4 | 389 | 92 | |
XM_005269115.2 | XP_005269172.1 | 3 - 4 | 396 | 92 | |
XM_005269117.1 | XP_005269174.1 | 3 - 4 | 438 | 92 | |
XM_006719581.1 | XP_006719644.1 | 2 - 3 | 238 | 92 | |
XM_017019910.1 | XP_016875399.1 | 3 - 4 | 389 | 92 | |
GenBank mRNA | AK290737.1 | - | 3 - 4 | 392 | 92 |
AK309569.1 | - | 3 - 4 | 438 | 92 | |
BC005860.2 | - | 3 - 4 | 684 | 92 | |
BI771121.1 | - | 3 - 4 | 397 | 92 | |
BT007140.1 | - | 3 - 4 | 331 | 92 | |
M60801.1 | - | 3 - 4 | 322 | 92 | |
U08998.2 | - | 3 - 4 | 419 | 92 |