The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014727.2 | NP_055542.1 | 13 - 14 | 3886 | 57 |
XM_011527561.2 | XP_011525863.2 | 13 - 14 | 3757 | 57 | |
XM_011527562.2 | XP_011525864.1 | 13 - 14 | 4087 | 57 | |
XM_017027544.1 | XP_016883033.1 | 13 - 14 | 4087 | 57 | |
XM_017027545.1 | XP_016883034.1 | 12 - 13 | 3800 | 57 | |
XM_017027546.1 | XP_016883035.1 | 8 - 9 | 1008 | 57 | |
GenBank mRNA | AB002302.3 | - | 11 - 12 | 2986 | 57 |
AF104918.1 | - | 11 - 12 | 1900 | 57 | |
AF186605.1 | - | 13 - 14 | 3558 | 57 | |
AJ007041.1 | - | 13 - 14 | 3886 | 57 |