The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316902.1 | NP_001303831.1 | 5 - 6 | 440 | 113 |
NM_014383.2 | NP_055198.1 | 5 - 6 | 1380 | 113 | |
XM_011526718.2 | XP_011525020.1 | 4 - 5 | 1541 | 113 | |
XM_017026589.1 | XP_016882078.1 | 5 - 6 | 1751 | 113 | |
GenBank mRNA | AF130255.1 | - | 4 - 5 | 1232 | 113 |
AF165097.1 | - | 3 - 4 | 1022 | 113 | |
HY039545.1 | - | 5 - 6 | 355 | 113 |