Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
Chromosome Location
Chr.12: 81078018 - 81257795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024560.2 NP_078836.1 7 - 8 1192 74
XM_005269150.2 XP_005269207.1 7 - 8 1126 74
XM_005269151.4 XP_005269208.1 7 - 8 1204 74
XM_011538740.2 XP_011537042.1 7 - 8 841 74
GenBank mRNA AK025616.1 - 7 - 8 1143 74
AK074938.1 - 2 - 3 373 74
AX764480.1 - 7 - 8 1342 74
AX764482.1 - 7 - 8 1098 74
BC009317.2 - 7 - 8 1192 74
BC015769.1 - 7 - 8 1192 74

View Full Product Details