Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSS3
Entrez Gene ID
79611
Gene Name
acyl-CoA synthetase short-chain family member 3
Gene Aliases
-
Chromosome Location
Chr.12: 81078018 - 81257795 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024560.2 NP_078836.1 8 - 9 1339 99
XM_005269150.2 XP_005269207.1 8 - 9 1273 99
XM_005269151.4 XP_005269208.1 8 - 9 1351 99
XM_011538740.2 XP_011537042.1 8 - 9 988 99
GenBank mRNA AK025616.1 - 8 - 9 1290 99
AX764480.1 - 8 - 9 1489 99
AX764482.1 - 8 - 9 1245 99
BC009317.2 - 8 - 9 1339 99
BC015769.1 - 8 - 9 1339 99

View Full Product Details