Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM160B2
Entrez Gene ID
64760
Gene Name
family with sequence similarity 160 member B2
Gene Aliases
RAI16
Chromosome Location
Chr.8: 22089150 - 22104380 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022749.5 NP_073586.5 4 - 5 453 107
XM_005273602.3 XP_005273659.1 4 - 5 453 107
XM_006716381.2 XP_006716444.1 4 - 5 453 107
XM_006716385.3 XP_006716448.1 5 - 6 1948 107
XM_017013732.1 XP_016869221.1 4 - 5 453 107
XM_017013733.1 XP_016869222.1 5 - 6 570 107
XM_017013734.1 XP_016869223.1 4 - 5 434 107
GenBank mRNA BC052237.1 - 4 - 5 450 107
CN307828.1 - 5 - 6 568 107

View Full Product Details