The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022749.5 | NP_073586.5 | 7 - 8 | 1017 | 78 |
XM_005273602.3 | XP_005273659.1 | 7 - 8 | 1017 | 78 | |
XM_006716381.2 | XP_006716444.1 | 7 - 8 | 1017 | 78 | |
XM_006716385.3 | XP_006716448.1 | 8 - 9 | 2512 | 78 | |
XM_017013732.1 | XP_016869221.1 | 7 - 8 | 1014 | 78 | |
XM_017013733.1 | XP_016869222.1 | 8 - 9 | 1134 | 78 | |
XM_017013734.1 | XP_016869223.1 | 7 - 8 | 998 | 78 | |
GenBank mRNA | AF495722.1 | - | 2 - 3 | 424 | 78 |
AK055191.1 | - | 1 - 2 | 76 | 78 | |
BC052237.1 | - | 7 - 8 | 1014 | 78 | |
BC112147.1 | - | 1 - 2 | 106 | 78 |