The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001304725.1 | NP_001291654.1 | 2 - 3 | 321 | 127 |
NM_017655.5 | NP_060125.4 | 2 - 3 | 618 | 127 | |
GenBank mRNA | AB073737.1 | - | 2 - 3 | 434 | 127 |
AK000082.1 | - | 1 - 2 | 177 | 127 | |
AK225941.1 | - | 2 - 3 | 457 | 127 | |
AK307422.1 | - | 2 - 3 | 321 | 127 | |
BC036075.1 | - | 2 - 3 | 618 | 127 |