Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
Chromosome Location
Chr.1: 2590639 - 2633042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033467.3 NP_258428.2 13 - 14 1490 65
XM_011542122.2 XP_011540424.1 6 - 7 593 65
XM_017002310.1 XP_016857799.1 14 - 15 1756 65
XM_017002311.1 XP_016857800.1 13 - 14 1436 65
XM_017002312.1 XP_016857801.1 12 - 13 1408 65
XM_017002313.1 XP_016857802.1 13 - 14 1490 65
XM_017002315.1 XP_016857804.1 13 - 14 1366 65
GenBank mRNA AF336981.1 - 12 - 13 1313 65
AK093058.1 - 13 - 14 1438 65
BC101027.2 - 12 - 13 1286 65
BC101028.1 - 8 - 9 815 65
BC101029.2 - 12 - 13 1286 65
BC101030.1 - 12 - 13 1286 65

View Full Product Details