Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
Chromosome Location
Chr.1: 2590639 - 2633042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033467.3 NP_258428.2 17 - 18 1907 98
XM_011542122.2 XP_011540424.1 10 - 11 1010 98
XM_017002310.1 XP_016857799.1 18 - 19 2173 98
XM_017002311.1 XP_016857800.1 17 - 18 1853 98
XM_017002312.1 XP_016857801.1 16 - 17 1825 98
XM_017002313.1 XP_016857802.1 17 - 18 1907 98
XM_017002314.1 XP_016857803.1 16 - 17 1778 98
XM_017002315.1 XP_016857804.1 17 - 18 1783 98
GenBank mRNA AF336981.1 - 16 - 17 1730 98
AK093058.1 - 17 - 18 1855 98
BC101027.2 - 16 - 17 1703 98
BC101028.1 - 12 - 13 1232 98
BC101030.1 - 16 - 17 1703 98

View Full Product Details