Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
Chromosome Location
Chr.1: 2590639 - 2633042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033467.3 NP_258428.2 22 - 23 2375 63
XM_011542122.2 XP_011540424.1 15 - 16 1478 63
XM_017002310.1 XP_016857799.1 23 - 24 2641 63
XM_017002311.1 XP_016857800.1 22 - 23 2321 63
XM_017002312.1 XP_016857801.1 21 - 22 2293 63
XM_017002314.1 XP_016857803.1 21 - 22 2246 63
XM_017002315.1 XP_016857804.1 22 - 23 2251 63
GenBank mRNA AF336981.1 - 21 - 22 2198 63
AK093058.1 - 22 - 23 2323 63
BC101027.2 - 21 - 22 2171 63
BC101028.1 - 17 - 18 1700 63
BC101029.2 - 21 - 22 2200 63
BC101030.1 - 21 - 22 2171 63

View Full Product Details