Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
Chromosome Location
Chr.1: 2590639 - 2633042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033467.3 NP_258428.2 5 - 6 670 56
XM_017002310.1 XP_016857799.1 6 - 7 936 56
XM_017002311.1 XP_016857800.1 5 - 6 670 56
XM_017002312.1 XP_016857801.1 4 - 5 588 56
XM_017002313.1 XP_016857802.1 5 - 6 670 56
XM_017002314.1 XP_016857803.1 5 - 6 670 56
XM_017002315.1 XP_016857804.1 5 - 6 546 56
GenBank mRNA AF336981.1 - 4 - 5 493 56
AK093058.1 - 5 - 6 618 56
BC101027.2 - 4 - 5 466 56
BC101029.2 - 4 - 5 466 56
BC101030.1 - 4 - 5 466 56

View Full Product Details