Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMEL1
Entrez Gene ID
79258
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
MMEL2, NEP2, NEPII, NL1, NL2, SEP
Chromosome Location
Chr.1: 2590639 - 2633042 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033467.3 NP_258428.2 6 - 7 742 64
XM_017002310.1 XP_016857799.1 7 - 8 1008 64
XM_017002311.1 XP_016857800.1 6 - 7 742 64
XM_017002312.1 XP_016857801.1 5 - 6 660 64
XM_017002313.1 XP_016857802.1 6 - 7 742 64
XM_017002314.1 XP_016857803.1 6 - 7 742 64
XM_017002315.1 XP_016857804.1 6 - 7 618 64
GenBank mRNA AF336981.1 - 5 - 6 565 64
AK093058.1 - 6 - 7 690 64
BC101027.2 - 5 - 6 538 64
BC101029.2 - 5 - 6 538 64
BC101030.1 - 5 - 6 538 64

View Full Product Details