The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301060.1 | NP_001287989.1 | 7 - 8 | 852 | 73 |
NM_024848.2 | NP_079124.1 | 7 - 8 | 852 | 73 | |
XM_005244798.2 | XP_005244855.1 | 6 - 7 | 716 | 73 | |
XM_011542169.1 | XP_011540471.1 | 7 - 8 | 852 | 73 | |
XM_011542170.2 | XP_011540472.1 | 7 - 8 | 852 | 73 | |
XM_011542172.2 | XP_011540474.1 | 6 - 7 | 716 | 73 | |
XM_011542173.2 | XP_011540475.1 | 7 - 8 | 852 | 73 | |
XM_011542174.2 | XP_011540476.1 | 7 - 8 | 852 | 73 | |
XM_011542176.1 | XP_011540478.1 | 7 - 8 | 852 | 73 | |
XM_011542177.1 | XP_011540479.1 | 7 - 8 | 852 | 73 | |
XM_017002365.1 | XP_016857854.1 | 7 - 8 | 852 | 73 | |
GenBank mRNA | AK024003.1 | - | 7 - 8 | 655 | 73 |
AK310323.1 | - | 6 - 7 | 716 | 73 | |
BC021704.2 | - | 7 - 8 | 808 | 73 |