The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203244.1 | NP_001190173.1 | 12 - 13 | 2030 | 75 |
NM_017893.3 | NP_060363.2 | 12 - 13 | 2030 | 75 | |
XM_005270008.2 | XP_005270065.1 | 13 - 14 | 2104 | 75 | |
GenBank mRNA | AB046839.1 | - | 12 - 13 | 1724 | 75 |
BC110382.1 | - | 12 - 13 | 1703 | 75 | |
BC128579.1 | - | 12 - 13 | 1847 | 75 |