The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145547.1 | NP_001139019.1 | 5 - 6 | 1155 | 147 |
NM_032905.4 | NP_116294.1 | 5 - 6 | 737 | 147 | |
GenBank mRNA | AF542550.1 | - | 5 - 6 | 679 | 147 |
AK314952.1 | - | 5 - 6 | 661 | 147 | |
BC007871.2 | - | 5 - 6 | 649 | 147 | |
BC009064.1 | - | 4 - 5 | 416 | 147 | |
BC039322.1 | - | 5 - 6 | 1158 | 147 |