The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001280796.1 | NP_001267725.1 | 8 - 9 | 1166 | 79 |
NM_001280797.1 | NP_001267726.1 | 7 - 8 | 1036 | 79 | |
NM_001280798.1 | NP_001267727.1 | 5 - 6 | 814 | 79 | |
NM_013267.3 | NP_037399.2 | 8 - 9 | 1166 | 79 | |
XM_005268797.1 | XP_005268854.1 | 8 - 9 | 793 | 79 | |
XM_017019180.1 | XP_016874669.1 | 7 - 8 | 663 | 79 | |
GenBank mRNA | AF110330.1 | - | 8 - 9 | 921 | 79 |
AF223944.1 | - | 8 - 9 | 1093 | 79 | |
AK000467.1 | - | 8 - 9 | 913 | 79 | |
BC025661.1 | - | 7 - 8 | 751 | 79 | |
DA060302.1 | - | 5 - 6 | 398 | 79 | |
HM775422.1 | - | 7 - 8 | 785 | 79 | |
HM775423.1 | - | 6 - 7 | 655 | 79 | |
HM775424.1 | - | 7 - 8 | 785 | 79 |