The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256053.1 | NP_001242982.1 | 20 - 21 | 2324 | 100 |
NM_014915.2 | NP_055730.2 | 20 - 21 | 2327 | 100 | |
XM_006717423.2 | XP_006717486.1 | 21 - 22 | 3413 | 100 | |
XM_006717425.3 | XP_006717488.1 | 21 - 22 | 3413 | 100 | |
XM_011519416.2 | XP_011517718.1 | 21 - 22 | 3413 | 100 | |
XM_017015928.1 | XP_016871417.1 | 21 - 22 | 3413 | 100 | |
XM_017015929.1 | XP_016871418.1 | 22 - 23 | 3401 | 100 | |
XM_017015930.1 | XP_016871419.1 | 21 - 22 | 3413 | 100 | |
XM_017015931.1 | XP_016871420.1 | 21 - 22 | 3413 | 100 | |
XM_017015932.1 | XP_016871421.1 | 21 - 22 | 3413 | 100 | |
XM_017015933.1 | XP_016871422.1 | 21 - 22 | 3413 | 100 | |
GenBank mRNA | AB028997.1 | - | 20 - 21 | 2302 | 100 |
AK128577.1 | - | 5 - 6 | 887 | 100 | |
BC130453.1 | - | 20 - 21 | 2268 | 100 |