The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006536.5 | NP_006527.1 | 7 - 8 | 1368 | 71 |
XM_011542448.2 | XP_011540750.1 | 7 - 8 | 1310 | 71 | |
GenBank mRNA | AB026833.1 | - | 7 - 8 | 1247 | 71 |
AF043977.1 | - | 7 - 8 | 1314 | 71 | |
AF127980.1 | - | 7 - 8 | 1206 | 71 | |
AK290348.1 | - | 7 - 8 | 1273 | 71 | |
BC041096.1 | - | 7 - 8 | 1269 | 71 |