The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006536.5 | NP_006527.1 | 8 - 9 | 1541 | 143 |
XM_011542448.2 | XP_011540750.1 | 8 - 9 | 1483 | 143 | |
GenBank mRNA | AB026833.1 | - | 8 - 9 | 1420 | 143 |
AF043977.1 | - | 8 - 9 | 1487 | 143 | |
AF127980.1 | - | 8 - 9 | 1379 | 143 | |
AK290348.1 | - | 8 - 9 | 1446 | 143 | |
AK307937.1 | - | 8 - 9 | 1444 | 143 | |
BC041096.1 | - | 8 - 9 | 1442 | 143 |