The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006536.5 | NP_006527.1 | 9 - 10 | 1649 | 112 |
XM_011542448.2 | XP_011540750.1 | 9 - 10 | 1591 | 112 | |
GenBank mRNA | AB026833.1 | - | 9 - 10 | 1528 | 112 |
AF043977.1 | - | 9 - 10 | 1595 | 112 | |
AF127980.1 | - | 9 - 10 | 1487 | 112 | |
AK290348.1 | - | 9 - 10 | 1554 | 112 | |
AK307937.1 | - | 9 - 10 | 1552 | 112 | |
BC041096.1 | - | 9 - 10 | 1550 | 112 |