The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182530.2 | NP_872336.2 | 7 - 8 | 1102 | 97 |
XM_011539052.2 | XP_011537354.1 | 6 - 7 | 1583 | 97 | |
XM_011539055.1 | XP_011537357.1 | 5 - 6 | 570 | 97 | |
XM_017018967.1 | XP_016874456.1 | 6 - 7 | 1384 | 97 | |
XM_017018968.1 | XP_016874457.1 | 7 - 8 | 1838 | 97 | |
XM_017018969.1 | XP_016874458.1 | 6 - 7 | 1318 | 97 | |
XM_017018970.1 | XP_016874459.1 | 5 - 6 | 1128 | 97 | |
XM_017018971.1 | XP_016874460.1 | 7 - 8 | 1907 | 97 | |
XM_017018972.1 | XP_016874461.1 | 7 - 8 | 1906 | 97 | |
XM_017018973.1 | XP_016874462.1 | 3 - 4 | 799 | 97 | |
XM_017018974.1 | XP_016874463.1 | 7 - 8 | 1907 | 97 | |
XM_017018975.1 | XP_016874464.1 | 7 - 8 | 1907 | 97 | |
XM_017018976.1 | XP_016874465.1 | 7 - 8 | 1907 | 97 |