Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYRFL
Entrez Gene ID
196446
Gene Name
myelin regulatory factor-like
Gene Aliases
C12orf15, C12orf28, bcm1377
Chromosome Location
Chr.12: 69824518 - 69958726 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182530.2 NP_872336.2 12 - 13 1687 75
XM_011539052.2 XP_011537354.1 11 - 12 2168 75
XM_011539055.1 XP_011537357.1 10 - 11 1155 75
XM_017018967.1 XP_016874456.1 11 - 12 1969 75
XM_017018968.1 XP_016874457.1 12 - 13 2423 75
XM_017018969.1 XP_016874458.1 11 - 12 1903 75
XM_017018970.1 XP_016874459.1 10 - 11 1713 75
XM_017018971.1 XP_016874460.1 12 - 13 2492 75
XM_017018972.1 XP_016874461.1 12 - 13 2491 75
XM_017018973.1 XP_016874462.1 8 - 9 1384 75
XM_017018974.1 XP_016874463.1 12 - 13 2492 75
XM_017018975.1 XP_016874464.1 12 - 13 2492 75
XM_017018976.1 XP_016874465.1 12 - 13 2492 75
GenBank mRNA EG328354.1 - 2 - 3 75

View Full Product Details