The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037984.2 | NP_001033073.1 | 2 - 3 | 637 | 96 |
NM_138570.3 | NP_612637.1 | 2 - 3 | 637 | 96 | |
XM_005257019.1 | XP_005257076.1 | 2 - 3 | 731 | 96 | |
GenBank mRNA | BC014642.1 | - | 2 - 3 | 564 | 96 |
BQ642219.1 | - | 2 - 3 | 604 | 96 |