Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 12 - 13 1565 125
XM_011543247.2 XP_011541549.1 12 - 13 1589 125
XM_011543248.2 XP_011541550.1 12 - 13 1709 125
XM_011543249.2 XP_011541551.1 7 - 8 1868 125
XM_017009174.1 XP_016864663.1 11 - 12 2324 125
XM_017009175.1 XP_016864664.1 8 - 9 2621 125
XM_017009176.1 XP_016864665.1 13 - 14 1737 125
GenBank mRNA AJ311904.1 - 13 - 14 2165 125
AK122622.1 - 3 - 4 411 125
AK128729.1 - 11 - 12 1380 125

View Full Product Details