Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADAMTS19
Entrez Gene ID
171019
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 19
Gene Aliases
-
Chromosome Location
Chr.5: 129460281 - 129738683 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011543246.2 XP_011541548.1 20 - 21 2726 62
XM_011543247.2 XP_011541549.1 20 - 21 2750 62
XM_011543248.2 XP_011541550.1 20 - 21 2870 62
XM_011543249.2 XP_011541551.1 15 - 16 3029 62
XM_017009174.1 XP_016864663.1 19 - 20 3485 62
XM_017009175.1 XP_016864664.1 16 - 17 3782 62
XM_017009176.1 XP_016864665.1 21 - 22 2898 62
GenBank mRNA AJ311904.1 - 21 - 22 3293 62
AK122622.1 - 11 - 12 1539 62
AK128729.1 - 20 - 21 2544 62

View Full Product Details