The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286568.1 | NP_001273497.1 | 13 - 14 | 2163 | 151 |
NM_001286569.1 | NP_001273498.1 | 14 - 15 | 2286 | 151 | |
NM_032143.3 | NP_115519.2 | 13 - 14 | 2068 | 151 | |
XM_005263809.1 | XP_005263866.1 | 13 - 14 | 2022 | 151 | |
XM_006712788.1 | XP_006712851.1 | 13 - 14 | 1996 | 151 | |
XM_011511958.2 | XP_011510260.1 | 13 - 14 | 2511 | 151 | |
XM_011511959.2 | XP_011510261.2 | 13 - 14 | 2076 | 151 | |
XM_011511960.1 | XP_011510262.1 | 13 - 14 | 2044 | 151 | |
XM_011511961.2 | XP_011510263.2 | 13 - 14 | 2010 | 151 | |
XM_011511963.1 | XP_011510265.1 | 13 - 14 | 2015 | 151 | |
XM_011511966.2 | XP_011510268.2 | 11 - 12 | 1894 | 151 | |
XM_017005065.1 | XP_016860554.1 | 13 - 14 | 2015 | 151 | |
XM_017005066.1 | XP_016860555.1 | 13 - 14 | 2074 | 151 | |
XM_017005067.1 | XP_016860556.1 | 13 - 14 | 2022 | 151 | |
XM_017005068.1 | XP_016860557.1 | 13 - 14 | 1996 | 151 | |
XM_017005069.1 | XP_016860558.1 | 13 - 14 | 2044 | 151 | |
XM_017005070.1 | XP_016860559.1 | 13 - 14 | 2044 | 151 | |
XM_017005071.1 | XP_016860560.1 | 11 - 12 | 1815 | 151 | |
GenBank mRNA | AK095362.1 | - | 12 - 13 | 1726 | 151 |
AK131303.1 | - | 7 - 8 | 1212 | 151 | |
AK304601.1 | - | 1 - 2 | 403 | 151 | |
AL136824.1 | - | 2 - 3 | 510 | 151 | |
AL833322.1 | - | 14 - 15 | 2167 | 151 | |
BC089429.1 | - | 13 - 14 | 2068 | 151 | |
BX647838.1 | - | 13 - 14 | 2191 | 151 |