The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288587.2 | NP_001275516.1 | 3 - 4 | 1380 | 88 |
GenBank mRNA | BC007828.2 | - | 3 - 4 | 1051 | 88 |
DQ280322.1 | - | 3 - 4 | 1090 | 88 |