The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012368.2 | NP_036500.2 | 1 - 1 | 792 | 85 |
GenBank mRNA | BC069158.1 | - | 1 - 1 | 740 | 85 |
BC069456.1 | - | 1 - 1 | 740 | 85 | |
BC126269.1 | - | 1 - 1 | 792 | 85 | |
BC130328.1 | - | 1 - 1 | 792 | 85 |